missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C-type lectin domain family 4 member C Protein

A DNA sequence encoding the Homo sapiens (Human) C-type lectin domain family 4 member C, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7991-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052031

  • 253.00€ / 50 microgramos


Fecha estimada de envío
19-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_569708.1
C-type lectin domain family 4 member C Protein
50 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
170482
Greater than 80% as determined by SDS-PAGE.
Research Use Only
CLEC4C
Recombinant Protein
E. coli
NFMYSKTVKR LSKLREYQQY HPSLTCVMEG KDIEDWSCCP TPWTSFQSSC YFISTGMQSW TKSQKNCSVM GADLVVINTR EEQDFIIQNL KRNSSYFLGL SDPGGRRHWQ WVDQTPYNEN VTFWHSGEPN NLDERCAIIN FRSSEEWGWN DIHCHVPQKS ICKMKKIYI
Videos
Documentos