missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C-Myc-binding Protein

A DNA sequence encoding the Homo sapiens (Human) C-Myc-binding protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8016-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052171

  • 160.00€ / 10 microgramos


Fecha estimada de envío
11-05-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_036465.2
C-Myc-binding Protein
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
26292
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MYCBP
Recombinant Protein
E. coli
AHYKAADSKR EQFRRYLEKS GVLDTLTKVL VALYEEPEKP NSALDFLKHH LGAATPENPE IELLRLELAE MKEKYEAIVE ENKKLKAKLA QYEPPQEEKR AE
Videos
Documentos