missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C-C motif chemokine 25 Protein

A DNA sequence encoding the Homo sapiens (Human) C-C motif chemokine 25, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5774-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937669

  • 406.00€ / 100 microgramos


Fecha estimada de envío
27-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001188288.1
C-C motif chemokine 25 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6370
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CCL25
Recombinant Protein
E. coli
QGVFEDCCLA YHYPIGWAVL RRAWTYRIQE VSGSCNLPAA IFYLPKRHRK VCGNPKSREV QRAMKLLDAR NKVFAKLHHN TQTFQAGPHA VKKLSSGNSK LSSSKFSNPI SSSKRNVSLL ISANSGL
Videos
Documentos