missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BUD31 Protein

A cDNA sequence encoding the BUD31 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11213-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916508

  • 103.00€ / 2 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

8896
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
BUD31
Human
His
BUD31 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT.
BUD31 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPKVKRS RKAPPDGWEL IEPTLDELDQ KMREAETEPH EGKRKVESLW PIFRIHHQKT RYIFDLFYKR KAISRELYEY CIKEGYADKN LIAKWKKQGY ENLCCLRCIQ TRDTNFGTNC ICRVPKSKLE VGRIIECTHC GCRGCSG
Videos