missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BUD31 Protein
A cDNA sequence encoding the BUD31 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11213-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
8896 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPKVKRS RKAPPDGWEL IEPTLDELDQ KMREAETEPH EGKRKVESLW PIFRIHHQKT RYIFDLFYKR KAISRELYEY CIKEGYADKN LIAKWKKQGY ENLCCLRCIQ TRDTNFGTNC ICRVPKSKLE VGRIIECTHC GCRGCSG | |
Greater than 85.0% as determined by SDS-PAGE. |
BUD31 Protein | |
Research Use Only | |
BUD31 | |
Human | |
His | |
BUD31 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT. |