missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BTC / Betacellulin Protein

A DNA sequence encoding the Human BTC / Betacellulin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10416-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913788

  • 668.00€ / 100 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001303892.1
BTC / Betacellulin Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
DGNSTRSPET NGLLCGDPEE NCAATTTQSK RKGHFSRCPK QYKHYCIKGR CRFVVAEQTP SCVCDEGYIG ARCERVDLFY
685
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
BTC
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 0.05 ng/ml, corresponding to a specific activity of > 2.0 M 107 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, with 0.02 % Tween-20, lyophilized
Videos