missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Breast cancer metastasis-suppressor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Breast cancer metastasis-suppressor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8482-ec-200ug

Additional Details : Peso : 0.01000kg

 View more versions of this product

Product Code. 16054861

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Description

Description

Specifications

Specifications

NP_056214.1
Breast cancer metastasis-suppressor 1 Protein
Research Use Only
BRMS1
Recombinant Protein
E. coli
MPVQPPSKDT EEMEAEGDSA AEMNGEEEES EEERSGSQTE SEEESSEMDD EDYERRRSEC VSEMLDLEKQ FSELKEKLFR ERLSQLRLRL EEVGAERAPE YTEPLGGLQR SLKIRIQVAG IYKGFCLDVI RNKYECELQG AKQHLESEKL LLYDTLQGEL QERIQRLEED RQSLDLSSEW WDDKLHARGS SRSWDSLPPS KRKKAPLVSG PYIVYMLQEI DILEDWTAIK KARAA
Greater than 90% as determined by SDS-PAGE.
25855
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers