missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Breast cancer metastasis-suppressor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Breast cancer metastasis-suppressor 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8482-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16034861

  • 160.00€ / 10 microgramos


Fecha estimada de envío
06-07-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_056214.1
Breast cancer metastasis-suppressor 1 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
25855
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BRMS1
Recombinant Protein
E. coli
MPVQPPSKDT EEMEAEGDSA AEMNGEEEES EEERSGSQTE SEEESSEMDD EDYERRRSEC VSEMLDLEKQ FSELKEKLFR ERLSQLRLRL EEVGAERAPE YTEPLGGLQR SLKIRIQVAG IYKGFCLDVI RNKYECELQG AKQHLESEKL LLYDTLQGEL QERIQRLEED RQSLDLSSEW WDDKLHARGS SRSWDSLPPS KRKKAPLVSG PYIVYMLQEI DILEDWTAIK KARAA
Videos
Documentos