missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRD2 Protein
A cDNA sequence encoding the BRD2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11203-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
BRD2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLQNVTP HNKLPGEGNA GLLGLGPEAA APGKRIRKPS LLYEGFESPT MASVPALQLT PANPPPPEVS NPKKPGRVTN QLQYLHKVVM KALWKHQFAW PFRQPVDAVK LGLPDYHKII KQPMDMGTIK RRLENNYYWA ASECMQDFNT MFTNCYIYNK PTDDIVLMAQ TLEKIFLQKV ASMPQEEQEL VVTIPKNSHK KGAKLAALQG SVTSAHQVPA VSSVSHTALY TPPPEIPTTV LNIPHPSVIS SPLLKSLHSA GPPLLAVTAA PPAQPLAKKK GVKRKADTTT PTPTAILAPG SPASPPGSLE PKAARLPPMR RESGRPIKPP RKDLPDSQQQ HQSSKKGKLS EQLKHCNGIL KELLSKKHAA YAWPFYKPVD ASALGLHDYH DIIKHPMDLS TVKRKMENRD YRDAQEFAAD VRLMFSNCYK YNPPDHDVVA MARKLQDVFE FRYAKMPD | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BRD2 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |