missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bone morphogenetic protein 1

A DNA sequence encoding the Homo sapiens (Human) Bone morphogenetic protein 3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5723-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937279

  • 675.00€ / 200 microgramos


Fecha estimada de envío
19-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001192.3
Bone morphogenetic protein 1
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
651
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BMP3
Recombinant Protein
E. coli
QWIEPRNCAR RYLKVDFADI GWSEWIISPK SFDAYYCSGA CQFPMPKSLK PSNHATIQSI VRAVGVVPGI PEPCCVPEKM SSLSILFFDE NKNVVLKVYP NMTVESCACR
Videos
Documentos