missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bone morphogenetic protein 1

A DNA sequence encoding the Homo sapiens (Human) Bone morphogenetic protein 7, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8515-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055061

  • 253.00€ / 50 microgramos


Fecha estimada de envío
16-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001710.1
Bone morphogenetic protein 1
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
655
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BMP7
Recombinant Protein
E. coli
STGSKQRSQN RSKTPKNQEA LRMANVAENS SSDQRQACKK HELYVSFRDL GWQDWIIAPE GYAAYYCEGE CAFPLNSYMN ATNHAIVQTL VHFINPETVP KPCCAPTQLN AISVLYFDDS SNVILKKYRN MVVRACGCH
Videos
Documentos