missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bone marrow proteoglycan Protein

A DNA sequence encoding the Homo sapiens(Human) Bone marrow proteoglycan, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9345-iv-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16058131

  • 910.00€ / 50 microgramos


Fecha estimada de envío
22-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001230174.1
Bone marrow proteoglycan Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
5553
Greater than 85% as determined by SDS-PAGE.
Research Use Only
PRG2
Recombinant Protein
Invitro-E. coli
TCRYLLVRSL QTFSQAWFTC RRCYRGNLVS IHNFNINYRI QCSVSALNQG QVWIGGRITG SGRCRRFQWV DGSRWNFAYW AAHQPWSRGG HCVALCTRGG HWRRAHCLRR LPFICSY
Videos
Documentos