missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BLVRB Protein
A cDNA sequence encoding the BLVRB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11170-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
BLVRB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAVKKIAIFG ATGQTGLTTL AQAVQAGYEV TVLVRDSSRL PSEGPRPAHV VVGDVLQAAD VDKTVAGQDA VIVLLGTRND LSPTTVMSEG ARNIVAAMKA HGVDKVVACT SAFLLWDPTK VPPRLQAVTD DHIRMHKVLR ESGLKYVAVM PPHIGDQPLT GAYTVTLDGR GPSRVISKHD LGHFMLRCLT TDEYDGHSTY PSHQYQ | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein contains 20mM Tris-HCl buffer pH 8.5, 10% glycerol, and 1mM DTT. |