missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BLK Protein
A cDNA sequence encoding the BLK was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11166-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
BLK Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGLVSSKKPD KEKPIKEKDK GQWSPLKVSA QDKDAPPLPP LVVFNHLTPP PPDEHLDEDK HFVVALYDYT AMNDRDLQML KGEKLQVLKG TGDWWLARSL VTGREGYVPS NFVARVESLE MERWFFRSQG RKEAERQLLA PINKAGSFLI RESETNKGAFSLSVKDVTTQ GELIKHYKIR CLDEGGYYIS PRITFPSLQA LVQHYSKKGD GLCQRLTLPC VRPAPQNPWA QDEWEIPRQS LRLVRKLGSG QFGEVWMGYY KNNMKVAIKT LKEGTMSPEA FLGEANVMKA LQHERLVRLY AVVTKEPIYI VTEYMARGCL LDFLKTDEGS RLSLPRLIDM SAQIAEGMAY IERMNSIHRD LRAANILVSE ALCCKIADFG LARIIDSEYT AQEGAKFPIK WTAPEAIHFG VFTIKADVWS FGVLLMEVVT YGRVPYPGMS NPEVIRNLER GYRMPRPDTC PPELYRGVIA ECWRSRPEER PTFEFLQSVL EDFYTATERQ YELQP | |
Greater than 80.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BLK protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.1M NaCl, 1mM DTT and 0.1mM PMSF. |