missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Biliverdin reductase A Protein

A DNA sequence encoding the Homo sapiens (Human) Biliverdin reductase A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5721-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937269

  • 234.00€ / 50 microgramos


Fecha estimada de envío
18-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000703.2
Biliverdin reductase A Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
644
Greater than 80% as determined by SDS-PAGE.
Research Use Only
BLVRA
Recombinant Protein
E. coli
AEPERKFGVV VVGVGRAGSV RMRDLRNPHP SSAFLNLIGF VSRRELGSID GVQQISLEDA LSSQEVEVAY ICSESSSHED YIRQFLNAGK HVLVEYPMTL SLAAAQELWE LAEQKGKVLH EEHVELLMEE FAFLKKEVVG KDLLKGSLLF TAGPLEEERF GFPAFSGISR LTWLVSLFGE LSLVSATLEE RKEDQYMKMT VCLETEKKSP LSWIEEKGPG LKRNRYLSFH FKSGSLENVP NVGVNKNIFL KDQNIFVQKL LGQFSEKELA AEKKRILHCL GLAEEIQKYC CSRK
Videos
Documentos