missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Biliverdin reductase A Protein
A DNA sequence encoding the Homo sapiens (Human) Biliverdin reductase A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5721-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000703.2 | |
Biliverdin reductase A Protein | |
Research Use Only | |
BLVRA | |
Recombinant Protein | |
E. coli | |
AEPERKFGVV VVGVGRAGSV RMRDLRNPHP SSAFLNLIGF VSRRELGSID GVQQISLEDA LSSQEVEVAY ICSESSSHED YIRQFLNAGK HVLVEYPMTL SLAAAQELWE LAEQKGKVLH EEHVELLMEE FAFLKKEVVG KDLLKGSLLF TAGPLEEERF GFPAFSGISR LTWLVSLFGE LSLVSATLEE RKEDQYMKMT VCLETEKKSP LSWIEEKGPG LKRNRYLSFH FKSGSLENVP NVGVNKNIFL KDQNIFVQKL LGQFSEKELA AEKKRILHCL GLAEEIQKYC CSRK | |
Greater than 80% as determined by SDS-PAGE. |
644 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |