Learn More
R&D Systems™ Recombinant Human Betacellulin Protein, Animal-Free
Description
Measured in a cell proliferation assay using Balb/3T3 mouse embryonic fibroblast cells. Rubin, J.S. et al. (1991) Proc. Natl. Acad. Sci. USA 88:415. The ED50 for this effect is 0.100-1.50 ng/mL.
Specifications
Specifications
| Accession Number | P35070.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 685 |
| Molecular Weight (g/mol) | M.W.-theroretical: 9.9 kDa |
| Quantity | 1 mg |
| Source | E. coli-derived human Betacellulin/BTC protein Asp32-Gln118, with an N-terminal Met Produced using non-animal reagents in an animal-free laboratory. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | betacellulin, BTC, probetacellulin |
| Conjugate | Unconjugated |
| Show More |