missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human beta-NGF / Beta-NGF Protein

Recombinant Protein

Marca:  enQuireBio™ QP5351-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915959

  • 91.15€ / 5 microgramos


Fecha estimada de envío
14-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_002497.2
beta-NGF / Beta-NGF Protein
5 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 5 ng/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
4803
Greater than 95% as determined by SDS-PAGE.
Research Use Only
NGF
Recombinant Protein
E. coli
MSSSHPIFHR GEFSVCDSVS VWVGDKTTAT DIKGKEVMVL GEVNINNSVF KQYFFETKCR DPNPVDSGCR GIDSKHWNSY CTTTHTFVKA LTMDGKQAAW RFIRIDTACV CVLSRKAVRR A
Videos
Documentos