missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta-defensin 4A Protein
A DNA sequence encoding the Homo sapiens (Human) Beta-defensin 4A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5927-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001192195.1 | |
Beta-defensin 4A Protein | |
Research Use Only | |
DEFB4A | |
Recombinant Protein | |
E. coli | |
GIGDPVTCLK SGAICHPVFC PRRYKQIGTC GLPGTKCCKK P | |
Greater than 80% as determined by SDS-PAGE. |
100289462 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |