missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Beta Defensin 103AB Protein

A DNA sequence encoding the Human Beta Defensin 103AB, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10431-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933838

  • 1970.00€ / 250 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001075020.1
Beta Defensin 103AB Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
GIINTLQKYY CRVRGGRCAV LSCLPKEEQI GKCSTRGRKC CRRKK
414325
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
DEFB103AB
The ED50 as determined by anti-microbial activity against E.coli. is less than 30 ug/ml, corresponding to a specific activity of > 33.3 IU/mg.
Human
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 130 mM NaCl, lyophilized
Videos
Documentos