missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BDNF Protein

A DNA sequence encoding the Homo sapiens (Human) BDNF Protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8513-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055051

  • 160.00€ / 10 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001137277.1
BDNF Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
627
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BDNF
Recombinant Protein
E. coli
ELSVCDSISE WVTAADKKTA VDMSGGTVTV LEKVPVSKGQ LKQYFYETKC NPMGYTKEGC RGIDKRHWNS QCRTTQSYVR ALTMDSKKRI GWRFIRIDTS C
Videos
Documentos