missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bcl 2 (minus BH3 domain) Protein

A cDNA sequence encoding the Bcl 2 (minus BH3 domain) was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10493-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924038

Cantidad Precio
1 1545.00€ / 500 microgramos
3 1441.49€ / 500 microgramos
Fecha estimada de envío
09-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

Bcl 2 (minus BH3 domain) Protein
500 μg
Untested
Yellow fever virus
His
>95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Human
DPBS pH 7.4.
Videos
Documentos