missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BCAT2 Protein

A DNA sequence encoding the Homo sapiens (Human) BCAT2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5707-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917169

  • 253.00€ / 50 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001158245.1
BCAT2 Protein
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
587
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BCAT2
Recombinant Protein
E. coli
ASSSFKAADL QLEMTQKPHK KPGPGEPLVF GKTFTDHMLM VEWNDKGWGQ PRIQPFQNLT LHPASSSLHY SLQLFEGMKA FKGKDQQVRL FRPWLNMDRM LRSAMRLCLP SFDKLELLEC IRRLIEVDKD WVPDAAGTSL YVRPVLIGNE PSLGVSQPTR ALLFVILCPV GAYFPGGSVT PVSLLADPAF IRAWVGGVGN YKLGGNYGPT VLVQQEALKR GCEQVLWLYG PDHQLTEVGT MNIFVYWTHE DGVLELVTPP LNGVILPGVV RQSLLDMAQT WGEFRVVERT ITMKQLLRAL EEGRVREVFG SGTACQVCPV HRILYKDRNL HIPTMENGPE LILRFQKELK EIQYGIRAHE WMFPV
Videos