missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BAFFR / TNFRSF13C Protein

A DNA sequence encoding the Human BAFFR / TNFRSF13C, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10356-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913608

  • 587.00€ / 250 microgramos


Fecha de envío estimada
del proveedor 22-05-2019
Añadir a la cesta

Especificación

Especificación

NP_443177.1
BAFFR / TNFRSF13C Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MRRGPRSLRG RDAPAPTPCV PAECFDLLVR HCVACGLLRT PRPKPAGASS PAPRTALQPQ ESVGAGAGEA ALPLPG
115650
Greater than 95% as determined by SDS-PAGE.
Research Use Only
TNFRSF13C
The ED50 as determined by its ability to block BAFF induced mouse splenocyte survival is 1.0-5.0 ug/ml in the presence of 1.0 ug/ml of rHuBAFF.
Human
Untagged
0.2 um filtered 20 mM PBS, pH 8.0, 500 mM NaCl, lyophilized
Videos