missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BAD Protein
A cDNA sequence encoding the BAD Protein was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11123-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
572 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MFQIPEFEPS EQEDSSSAER GLGPSPAGDG PSGSGKHHRQ APGLLWDASH QQEQPTSSSH HGGAGAVEIR SRHSSYPAGT EDDEGMGEEP SPFRGRSRSA PPNLWAAQRY GRELRRMSDE FVDSFKKGLP RPKSAGTATQ MRQSSSWTRV FQSWWDRNLG RGSSAPSQ |
BAD Protein | |
Research Use Only | |
BAD | |
Human | |
GST | |
BAD protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA, 20% Glycerol. |