missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human B2M Protein

A cDNA sequence encoding the B2M was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11117-HIS-1ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916208

  • 103.00€ / 1 microgramo


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

567
Greater than 98.0%.
Research Use Only
B2M
Human
His
Lyophilized from 0.02M NH4HCO3.
B2M Protein
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MSRSVALAVL ALLSLSGLEA IQRTPKIQVY SRHPAENGKS NFLNCYVSGF HPSDIEVDLL KNGERIEKVE HSDLSFSKDW SFYLLYYTEF TPTEKDEYAC RVNHVTLSQP KIVKWDRDM
Videos