missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Autophagy-related protein 16-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Autophagy-related protein 16-1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7727-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050451

  • 1855.00€ / 1 miligramo


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001177195.1
Autophagy-related protein 16-1 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
55054
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ATG16L1
Recombinant Protein
E. coli
MSSGLRAADF PRWKRHISEQ LRRRDRLQRQ AFEEIILQYN KLLEKSDLHS VLAQKLQAEK HDVPNRHEIS PGHDGTWNDN QLQEMAQLRI KHQEELTELH KKRGELAQLV IDLNNQMQRK DREMQMNEAK IAECLQTISD LETECLDLRT KLCDLERANQ TLKDEYDALQ ITFTALEGKL RKTTEENQEL VTRWMAEKAQ EANRLNAENE KDSRRRQARL QKELAEAAKE PLPVEQDDDI EVIVDETSDH TEETSPVRAI SRAATKRLSQ PAGGLLDSIT NIFGRRSVSS FPVPQDNVDT HPGSGKEVRV PATALCVFDA HDGEVNAVQF SPGSRLLATG GMDRRVKLWE VFGEKCEFKG SLSGSNAGIT SIEFDSAGSY LLAASNDFAS RIWTVDDYRL RHTLTGHSGK VLSAKFLLDN ARIVSGSHDR TLKLWDLRSK VCIKTVFAGS SCNDIVCTEQ CVMSGHFDKK IRFWDIRSES IVREMELLGK ITALDLNPER TELLSCSRDD LLKVIDLRTN AIKQTFSAPG FKCGSDWTRV VFSPDGSYVA AGSAEGSLYI WSVLTGKVEK VLSKQHSSSI NAVAWSPSGS HVVSVDKGCK AVLWAQY
Videos
Documentos