missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Atypical chemokine receptor 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Atypical chemokine receptor 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5898-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_064707.1 | |
Atypical chemokine receptor 3 Protein | |
Research Use Only | |
ACKR3 | |
Recombinant Protein | |
E. coli | |
MDLHLFDYSE PGNFSDISWP CNSSDCIVVD TVMCPNMPNK | |
Greater than 90% as determined by SDS-PAGE. |
57007 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |