missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP-dependent RNA helicase DDX19A Protein

A DNA sequence encoding the Homo sapiens (Human) ATP-dependent RNA helicase DDX19A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8225-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053391

  • 675.00€ / 200 microgramos


Fecha estimada de envío
20-01-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001307451.1
ATP-dependent RNA helicase DDX19A Protein
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
55308
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DDX19A
Recombinant Protein
E. coli
MATDSWALAV DEQEAAVKSM TNLQIKEEKV KADTNGIIKT STTAEKTDEE EKEDRAAQSL LNKLIRSNLV DNTNQVEVLQ RDPNSPLYSV KSFEELRLKP QLLQGVYAMG FNRPSKIQEN ALPMMLAEPP QNLIAQSQSG TGKTAAFVLA MLSRVEPSDR YPQCLCLSPT YELALQTGKV IEQMGKFYPE LKLAYAVRGN KLERGQKISE QIVIGTPGTV LDWCSKLKFI DPKKIKVFVL DEADVMIATQ GHQDQSIRIQ RMLPRNCQML LFSATFEDSV WKFAQKVVPD PNVIKLKREE ETLDTIKQYY VLCSSRDEKF QALCNLYGAI TIAQAMIFCH TRKTASWLAA ELSKEGHQVA LLSGEMMVEQ RAAVIERFRE GKEKVLVTTN VCARGIDVEQ VSVVINFDLP VDKDGNPDNE TYLHRIGRTG RFGKRGLAVN MVDSKHSMNI LNRIQEHFNK KIERLDTDDL DEIEKIAN
Videos
Documentos