missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP-dependent RNA helicase A Protein
A DNA sequence encoding the Homo sapiens (Human) ATP-dependent RNA helicase A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7512-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001348.2 | |
ATP-dependent RNA helicase A Protein | |
Research Use Only | |
DHX9 | |
Recombinant Protein | |
E. coli | |
NQVGVVPWSP PQSNWNPWTS SNIDEGPLAF ATPEQISMDL KNELMYQLEQ DHDLQAILQE RELLPVKKFE SEILEAISQN SVVIIRGATG CGKTTQVPQF ILDDFIQNDR AAECNIVVTQ PRRISAVSVA ERVAFERGEE PGKSCGYSVR FESILPRPHA SIMFCTVGVL LRKLEAGIRG ISHVIVDEIH ERDINTDFLL VVLRDVVQAY PEVRIVLMSA TIDTSMFCEY FFNCPIIEVY GRTYPVQEYF LEDCIQMTHF VPPPKDKKKK DKDDDGGEDD DANCNLICGD EYGPETRLSM SQLNEKETPF ELIEALLKYI ETLNVPGAVL VFLPGWNLIY TMQKHLEMNP HFGSHRYQIL PLHSQIPREE QRKVFDPVPV GVTKVILSTN IAETSITIND VVYVIDSCKQ KVKLFTAHNN MTNYATVWAS KTNLEQRKGR AGRVRPGFCF HLCSRARFER LETHMTPEMF RTPLHEIALS IKLLRLGGIG QFLAKAIEPP PLDAVIEAEH TLRELD | |
Greater than 90% as determined by SDS-PAGE. |
1660 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |