Learn More
enQuireBio™ Recombinant Human ASGPR1 / ASGR1 Protein
A DNA sequence encoding the Homo sapiens (Human) ASGPR1 / ASGR1, was expressed in the hosts and tags indicated.
Marca:  enQuireBio™ QP8899-ec-10ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 16087071
  • 212.00€ / 10 microgramos
Fecha estimada de envío: 28-05-2024
para ver el stock
Especificaciones
NP_001184145.1
ASGPR1 / ASGR1 Protein
Research Use Only
ASGR1
Recombinant Protein
E. coli
EELRGLRETF SNFTASTEAQ VKGLSTQGGN VGRKMKSLES QLEKQQKDLS EDHSSLLLHV KQFVSDLRSL SCQMAALQGN GSERTCCPVN WVEHERSCYW FSRSGKAWAD ADNYCRLEDA HLVVVTSWEE QKFVQHHIGP VNTWMGLHDQ NGPWKWVDGT DYETGFKNWR PEQPDDWYGH GLGGGEDCAH FTDDGRWNDD VCQRPYRWVC ETELDKASQE PPLL
Greater than 90% as determined by SDS-PAGE.
432
10 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol