missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ASB8 Protein

A cDNA sequence encoding the ASB8 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11082-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916088

  • 1372.00€ / 100 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

140461
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
ASB8
Human
His
The ASB8 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol.
ASB8 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSSSMWY IMQSIQSKYS LSERLIRTIA AIRSFPHDNV EDLIRGGADV NCTHGTLKPL HCACMVSDAD CVELLLEKGA EVNALDGYNR TALHYAAEKD EACVEVLLEY GANPNALDGN RDTPLHWAAF KNNAECVRAL LESGASVNAL DYNNDTPLSW AAMKGNLESV SILLDYGAEV RVINLIGQTP ISRLVALLVR GLGTEKEDSC FELLHRAVGH FELRKNGTMP REVARDPQLC EKLTVLCSAP GTLKTLARYA VRRSLGLQYL PDAVKGLPLP ASLKEYLLLL E
Videos