Learn More
enQuireBio™ Recombinant Human Artemin Protein
A cDNA sequence encoding the Artemin was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Especificaciones
Nombre Artemin Protein
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Actividad biológica The activity is determined by the dose-dependent proliferation of the SH-SY5Y cell line and is typically 4-8 ng/mL. The activity can also be determined by its ability to promote survival and neurite outgrowth.
Tipo de producto Recombinant Protein
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15924138
Ver documentos Detalles de la promoción
enQuireBio™
QP10524-5UG
5 μg
137.00€
5 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 29-05-2024
para ver el stock
15914138
Ver documentos Detalles de la promoción
enQuireBio™
QP10524-20UG
20 μg
212.00€
20 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 29-05-2024
para ver el stock
15904138
Ver documentos Detalles de la promoción
enQuireBio™
QP10524-1MG
1 mg
4090.00€
1 miligramo
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 29-05-2024
para ver el stock
Especificaciones
Artemin Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
AGGPGSRARA AGARGCRLRS QLVPVRALGL GHRSDELVRF RFCSGSCRRA RSPHDLSLAS LLGAGALRPP PGSRPVSQPC CRPTRYEAVS FMDVNSTWRT VDRLSATACG CLG
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
The activity is determined by the dose-dependent proliferation of the SH-SY5Y cell line and is typically 4-8 ng/mL. The activity can also be determined by its ability to promote survival and neurite outgrowth.
Human
Untagged
Artemin was lyophilized after extensive dialysis against 10mM sodium citrate pH 4.5 and 25mM sodium chloride.