missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Artemin Protein
A cDNA sequence encoding the Artemin was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10524-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Artemin Protein | |
Research Use Only | |
The activity is determined by the dose-dependent proliferation of the SH-SY5Y cell line and is typically 4-8 ng/mL. The activity can also be determined by its ability to promote survival and neurite outgrowth. | |
Human | |
Untagged | |
Artemin was lyophilized after extensive dialysis against 10mM sodium citrate pH 4.5 and 25mM sodium chloride. |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
AGGPGSRARA AGARGCRLRS QLVPVRALGL GHRSDELVRF RFCSGSCRRA RSPHDLSLAS LLGAGALRPP PGSRPVSQPC CRPTRYEAVS FMDVNSTWRT VDRLSATACG CLG | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |