missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Artemin Protein

A cDNA sequence encoding the Artemin was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10524-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914138

  • 212.00€ / 20 microgramos
Fecha estimada de envío: 28-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

Artemin Protein
Research Use Only
The activity is determined by the dose-dependent proliferation of the SH-SY5Y cell line and is typically 4-8 ng/mL. The activity can also be determined by its ability to promote survival and neurite outgrowth.
Human
Untagged
Artemin was lyophilized after extensive dialysis against 10mM sodium citrate pH 4.5 and 25mM sodium chloride.
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
AGGPGSRARA AGARGCRLRS QLVPVRALGL GHRSDELVRF RFCSGSCRRA RSPHDLSLAS LLGAGALRPP PGSRPVSQPC CRPTRYEAVS FMDVNSTWRT VDRLSATACG CLG
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales