missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human ART4 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

Marca:  Novus Biologicals™ NBP2-61454-10UG

Detalles adicionales : Peso : 0.00970kg

 Ver más versiones de este producto

Código de producto. 18215294

  • 346.64€ / 10 microgramos
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

A recombinant protein with a C-terminalHis-tag and corresponding to the amino acids 47-285 of Human ART4. The Recombinant Human ART4 Protein is derived from Baculovirus. The Recombinant Human ART4 Protein has been validated for the following applications: SDS-Page.
Especificaciones

Especificaciones

0.25 mg/ml
Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
28.8 kDa
Baculovirus
RUO
ART4
>85%, by SDS-PAGE
SDS-PAGE
420
10 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
ART4
Human
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales