missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ART1 / CD296 / ARTC1 Protein

A DNA sequence encoding the Homo sapiens (Human) ART1 / CD296 / ARTC1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5681-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936969

  • 277.00€ / 50 microgramos


Fecha estimada de envío
18-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_004305.2
ART1 / CD296 / ARTC1 Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
417
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ART1
Recombinant Protein
Yeast
QSHPITRRDL FSQEIQLDMA LASFDDQYAG CAAAMTAALP DLNHTEFQAN QVYADSWTLA SSQWQERQAR WPEWSLSPTR PSPPPLGFRD EHGVALLAYT ANSPLHKEFN AAVREAGRSR AHYLHHFSFK TLHFLLTEAL QLLGSGQRPP RCHQVFRGVH GLRFRPAGPR ATVRLGGFAS ASLKHVAAQQ FGEDTFFGIW TCLGAPIKGY SFFPGEEEVL IPPFETFQVI NASRLAQGPA RIYLRALGKH STYNCEYIKD KKCKSGPCHL DNS
Videos
Documentos