missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARL5B Protein
A cDNA sequence encoding the ARL5B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11074-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ARL5B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGLIFAKLWS LFCNQEHKVI IVGLDNAGKT TILYQFLMNE VVHTSPTIGS NVEEIVVKNT HFLMWDIGGQ ESLRSSWNTY YSNTEFIILV VDSIDRERLA ITKEELYRML AHEDLRKAAV LIFANKQDMK GCMTAAEISK YLTLSSIKDH PWHIQSCCAL TGEGLCQGLE WMTSRIGVR | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ARL5B protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 30% glycerol, 0.1M NaCl and 1mM EDTA. |