missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ARL11 Protein

A cDNA sequence encoding the ARL11 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11068-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916048

  • 103.00€ / 5 microgramos


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

ARL11 Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ARL11 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 5mM DTT,30% glycerol, 100mM NaCl and 1mM EDTA.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSVNSRGHK AEAQVVMMGL DSAGKTTLLY KLKGHQLVET LPTVGFNVEP LKAPGHVSLT LWDVGGQAPL RASWKDYLEG TDILVYVLDS TDEARLPESA AELTEVLNDP NMAGVPFLVL ANKQEAPDAL PLLKIRNRLS LERFQDHCWE LRGCSALTGE GLPEALQSLW SLLKSRSCMC LQARAHGAER GDSKRS
Videos
Documentos