missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ARF3 / ADP-ribosylation factor 3 Protein

A DNA sequence encoding the Homo sapiens (Human) ARF3 / ADP-ribosylation factor 3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5673-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916909

  • 726.00€ / 200 microgramos


Fecha estimada de envío
23-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001650.1
ARF3 / ADP-ribosylation factor 3 Protein
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
377
Greater than 80% as determined by SDS-PAGE.
Research Use Only
ARF3
Recombinant Protein
E. coli
GNIFGNLLKS LIGKKEMRIL MVGLDAAGKT TILYKLKLGE IVTTIPTIGF NVETVEYKNI SFTVWDVGGQ DKIRPLWRHY FQNTQGLIFV VDSNDRERVN EAREELMRML AEDELRDAVL LVFANKQDLP NAMNAAEITD KLGLHSLRHR NWYIQATCAT SGDGLYEGLD WLANQLKNKK
Videos