missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apoptosis regulatory protein Siva Protein
A DNA sequence encoding the Homo sapiens (Human) Apoptosis regulatory protein Siva, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6688-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006418.2 | |
Apoptosis regulatory protein Siva Protein | |
Research Use Only | |
SIVA1 | |
Recombinant Protein | |
E. coli | |
MPKRSCPFAD VAPLQLKVRV SQRELSRGVC AERYSQEVFD PSGVASIACS SCVRAVDGKA VCGQCERALC GQCVRTCWGC GSVACTLCGL VDCSDMYEKV LCTSCAMFET | |
Greater than 90% as determined by SDS-PAGE. |
10572 | |
1 mg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |