missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Apoptosis-inducing factor 1, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Apoptosis-inducing factor 1, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5633-ec-100ug

Additional Details : Peso : 0.01000kg

 View more versions of this product

Product Code. 15936589

  • 530.00€ / 100 microgramos
Fecha estimada de envío: 29-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Description

Description

Specifications

Specifications

NP_001124318.2
Apoptosis-inducing factor 1, mitochondrial Protein
Research Use Only
AIFM1
Recombinant Protein
E. coli
GLTPEQKQKK AALSASEGEE VPQDKAPSHV PFLLIGGGTA AFAAARSIRA RDPGARVLIV SEDPELPYMR PPLSKELWFS DDPNVTKTLR FKQWNGKERS IYFQPPSFYV SAQDLPHIEN GGVAVLTGKK VVQLDVRDNM VKLNDGSQIT YEKCLIATGG TPRSLSAIDR AGAEVKSRTT LFRKIGDFRS LEKISREVKS ITIIGGGFLG SELACALGRK ARALGTEVIQ LFPEKGNMGK ILPEYLSNWT MEKVRREGVK VMPNAIVQSV GVSSGKLLIK LKDGRKVETD HIVAAVGLEP NVELAKTGGL EIDSDFGGFR VNAELQARSN IWVAGDAACF YDIKLGRRRV EHHDHAVVSG RLAGENMTGA AKPYWHQSMF WSDLGPDVGY EAIGLVDSSL PTVGVFAKAT AQDNPKSATE QSGTGIRSES ETESEASEIT IPPSTPAVPQ APVQGEDYGK GVIFYLRDKV VVGIVLWNIF NRMPIARKII KDGEQHEDLN EVAKLFNIHE
Greater than 90% as determined by SDS-PAGE.
9131
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers