missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Apoptosis facilitator Bcl-2-like protein 14 Protein

A DNA sequence encoding the Homo sapiens (Human) Apoptosis facilitator Bcl-2-like protein 14, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8211-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053301

Cantidad Precio
1 985.00€ / 200 microgramos
4 919.50€ / 200 microgramos
Fecha estimada de envío
09-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_110393.1
Apoptosis facilitator Bcl-2-like protein 14 Protein
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
79370
Greater than 80% as determined by SDS-PAGE.
Research Use Only
BCL2L14
Recombinant Protein
E. coli
MCSTSGCDLE EIPLDDDDLN TIEFKILAYY TRHHVFKSTP ALFSPKLLRT RSLSQRGLGN CSANESWTEV SWPCRNSQSS EKAINLGKKK SSWKAFFGVV EKEDSQSTPA KVSAQGQRTL EYQDSHSQQW SRCLSNVEQC LEHEAVDPKV ISIANRVAEI VYSWPPPQAT QAGGFKSKEI FVTEGLSFQL QGHVPVASSS KKDEEEQILA KIVELLKYSG DQLERKLKKD KALMGHFQDG LSYSVFKTIT DQVLMGVDPR GESEVKAQGF KAALVIDVTA KLTAIDNHPM NRVLGFGTKY LKENFSPWIQ QHGGWEKILG ISHEEVD
Videos
Documentos