missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Apolipoprotein C-II Protein

A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein C-II, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5665-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936839

  • 287.00€ / 10 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000474.2
Apolipoprotein C-II Protein
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
344
Greater than 90% as determined by SDS-PAGE.
Research Use Only
APOC2
Recombinant Protein
Yeast
TQQPQQDEMP SPTFLTQVKE SLSSYWESAK TAAQNLYEKT YLPAVDEKLR DLYSKSTAAM STYTGIFTDQ VLSVLKGEE
Videos
Documentos