missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AP1AR Protein
A cDNA sequence encoding the AP1AR was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11038-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
55435 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGNCCWTQCF GLLRKEAGRL QRVGGGGGSK YFRTCSRGEH LTIEFENLVE SDEGESPGSS HRPLTEEEIV DLRERHYDSI AEKQKDLDKK IQKELALQEE KLRLEEEALY AAQREAARAA KQRKLLEQER QRIVQQYHPS NNGEYQSSGP EDDFESCLRN MKSQYEVFRS SRLSSDATVL TPNTESSCDL MTKTKSTSGN DDSTSLDLEW EDEEGMNRML PMRERSKTEE DILRAALKYS NKKTGSNPTS ASDDSNGLEW ENDFVSAEMD DNGNSEYSGF VNPVLELSDS GIRHSDTDQQ TR | |
Greater than 90.0% as determined by SDS-PAGE. |
AP1AR Protein | |
Research Use Only | |
AP1AR | |
Human | |
His | |
The AP1AR solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |