missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANXA10 Protein
A cDNA sequence encoding the ANXA10 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11031-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ANXA10 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMFCGDY VQGTIFPAPN FNPIMDAQML GGALQGFDCD KDMLINILTQ RCNAQRMMIA EAYQSMYGRD LIGDMREQLS DHFKDVMAGL MYPPPLYDAH ELWHAMKGVG TDENCLIEIL ASRTNGEIFQ MREAYCLQYS NNLQEDIYSE TSGHFRDTLMNLVQGTREEG YTDPAMAAQD AMVLWEACQQ KTGEHKTMLQ MILCNKSYQQ LRLVFQEFQN ISGQDMVDAI NECYDGYFQE LLVAIVLCVR DKPAYFAYRL YSAIHDFGFH NKTVIRILIA RSEIDLLTIR KRYKERYGKS LFHDIRNFAS GHYKKALLAI CAGDAEDY | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ANXA10 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.1M NaCl. |