missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Angiotensinogen / SerpinA8 / AGT Protein
A DNA sequence encoding the Homo sapiens (Human) Angiotensinogen / SerpinA8 / AGT, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5624-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000020.1 | |
Angiotensinogen / SerpinA8 / AGT Protein | |
Research Use Only | |
AGT | |
Recombinant Protein | |
E. coli | |
VIHNESTCEQ LAKANAGKPK DPTFIPAPIQ AKTSPVDEKA LQDQLVLVAA KLDTEDKLRA AMVGMLANFL GFRIYGMHSE LWGVVHGATV LSPTAVFGTL ASLYLGALDH TADRLQAILG VPWKDKNCTS RLDAHKVLSA LQAVQGLLVA QGRADSQAQL LLSTVVGVFT APGLHLKQPF VQGLALYTPV VLPRSLDFTE LDVAAEKIDR FMQAVTGWKT GCSLMGASVD STLAFNTYVH FQGKMKGFSL LAEPQEFWVD NSTSVSVPML SGMGTFQHWS DIQDNFSVTQ VPFTESACLL LIQPHYASDL DKVEGLTFQQ NSLNWMKKLS PRTIHLTMPQ LVLQGSYDLQ DLLAQAELPA ILHTELNLQK LSNDRIRVGE VLNS | |
Greater than 90% as determined by SDS-PAGE. |
183 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |