missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Angiopoietin-2 / ANG2 Protein

A DNA sequence encoding the Homo sapiens (Human) Angiopoietin-2 / ANG2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8512-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055041

  • 675.00€ / 200 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001112359.1
Angiopoietin-2 / ANG2 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
285
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ANGPT2
Recombinant Protein
E. coli
YNNFRKSMDS IGKKQYQVQH GSCSYTFLLP EMDNCRSSSS PYVSNAVQRD APLEYDDSVQ RLQVLENIME NNTQWLMKLE NYIQDNMKKE MVEIQQNAVQ NQTAVMIEIG TNLLNQTAEQ TRKLTDVEAQ VLNQTTRLEL QLLEHSLSTN KLEKQILDQT SEINKLQDKN SFLEKKVLAM EDKHIIQLQS IKEEKDQLQV LVSKQNSIIE ELEKKIVTAT VNNSVLQKQQ HDLMETVNNL LTMMSTSNSA KDPTVAKEEQ ISFRDCAEVF KSGHTTNGIY TLTFPNSTEE IKAYCDMEAG GGGWTIIQRR EDGSVDFQRT WKEYKVGFGN PSGEYWLGNE FVSQLTNQQR YVLKIHLKDW EGNEAYSLYE HFYLSSEELN YRIHLKGLTG TAGKISSISQ PGNDFSTKDG DNDKCICKCS QMLTGGWWFD ACGPSNLNGM YYPQRQNTNK FNGIKWYYWK GSGYSLK
Videos
Documentos