missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Anamorsin Protein
A DNA sequence encoding the Homo sapiens (Human) Anamorsin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7733-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001295276.1 | |
Anamorsin Protein | |
Research Use Only | |
CIAPIN1 | |
Recombinant Protein | |
E. coli | |
MADFGISAGQ FVAVVWDKSS PVEALKGLVD KLQALTGNEG RVSVENIKQL LQSAHKESSF DIILSGLVPG STTLHSAEIL AEIARILRPG GCLFLKEPVE TAVDNNSKVK TASKLCSALT LSGLVEVKEL QREPLTPEEV QSVREHLGHE SDNLLFVQIT GKKPNFEVGS SRQLKLSITK KSSPSVKPAV DPAAAKLWTL SANDMEDDSM DLIDSDELLD PEDLKKPDPA SLRAASCGEG KKRKACKNCT CGLAEELEKE KSREQMSSQP KSACGNCYLG DAFRCASCPY LGMPAFKPGE KVLLSDSNLH DA | |
Greater than 90% as determined by SDS-PAGE. |
57019 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |