missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Amphiregulin Protein
A DNA sequence encoding the Human Amphiregulin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10399-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001648.1 | |
Amphiregulin Protein | |
Research Use Only | |
AREG | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is between 5-10 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
374 | |
50 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SVRVEQVVKP PQNKTESENT SDKPKRKKKG GKNGKNRRNR KKKNPCNAEF QNFCIHGECK YIEHLEAVTC KCQQEYFGER CGEKSMKTHS MIDSSLSK | |
Greater than 95% as determined by SDS-PAGE and HPLC. |